Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
TC1900006522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145044.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586757.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000586843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000590136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006522.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
TC1900006522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
ENST00000591446.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145045.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145046.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145046.hg.1 |
NM_177401.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145046.hg.1 |
TC1900006522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145046.hg.1 |
ENST00000300952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145046.hg.1 |
NM_177401.4 |
Refseq |