Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005388.hg.1 |
ENST00000331159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005388.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005388.hg.1 |
TC17000460.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005386.hg.1 |
ENST00000331159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005386.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005386.hg.1 |
TC17000460.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005387.hg.1 |
ENST00000331159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005387.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005387.hg.1 |
TC17000460.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155538.hg.1 |
TC1700007702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007702.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155534.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155534.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155534.hg.1 |
TC1700007702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
ENST00000617765.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155540.hg.1 |
TC1700007702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
ENST00000617360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155537.hg.1 |
TC1700007702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
ENST00000612932.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
ENST00000613678.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
NM_014598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155536.hg.1 |
TC1700007702.hg.1 |
Clariom D TC |