Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018482.hg.1 |
TC07001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018482.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018482.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018482.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018482.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018478.hg.1 |
TC07001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018478.hg.1 |
ENST00000297210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018478.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018478.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018480.hg.1 |
TC07001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018480.hg.1 |
ENST00000297210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018480.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018480.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018480.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018480.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018487.hg.1 |
TC07001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018487.hg.1 |
ENST00000297210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018487.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018487.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018487.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018487.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018484.hg.1 |
TC07001183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018484.hg.1 |
ENST00000297210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018484.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018484.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018484.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07018484.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174108.hg.1 |
TC0700010412.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174116.hg.1 |
TC0700010412.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010412.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174106.hg.1 |
TC0700010412.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174110.hg.1 |
TC0700010412.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
ENST00000361443.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
ENST00000418710.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
ENST00000617581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
NM_182700.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
NM_198956.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700174112.hg.1 |
TC0700010412.hg.1 |
Clariom D TC |