Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144848.hg.1 |
TC1300006587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144849.hg.1 |
TC1300006587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300144852.hg.1 |
ENST00000443092.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144852.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144852.hg.1 |
ENST00000443092.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144852.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144852.hg.1 |
TC1300006587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006587.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144850.hg.1 |
TC1300006587.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
ENST00000443092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
NR_103450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300144851.hg.1 |
TC1300006587.hg.1 |
Clariom D TC |