Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022749.hg.1 |
TC03003375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022749.hg.1 |
ENST00000310232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022749.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022749.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022750.hg.1 |
TC03003375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022750.hg.1 |
ENST00000310232.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022750.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022750.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000466031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000466031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000466031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000472921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000472921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000472921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000466031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000466031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000466031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000472921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000472921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000472921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000479576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013959.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207144.hg.1 |
TC0300013959.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
ENST00000310232.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
ENST00000435327.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
ENST00000492806.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
NM_144719.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207146.hg.1 |
TC0300013959.hg.1 |
Clariom D TC |