Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017121.hg.1 |
ENST00000226218.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017121.hg.1 |
ENST00000536498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017121.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017121.hg.1 |
TC17001296.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205800.hg.1 |
TC1700012379.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
ENST00000542029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012379.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
ENST00000226218.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
ENST00000539746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
NM_000638.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205802.hg.1 |
TC1700012379.hg.1 |
Clariom D TC |