Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290223.1 | NP_001277152.1 | 40 - 41 | 4231 | 79 | |
NM_001380.4 | NP_001371.1 | 40 - 41 | 4168 | 79 | ||
XM_011539422.2 | XP_011537724.1 | 40 - 41 | 4209 | 79 | ||
XM_011539424.1 | XP_011537726.1 | 40 - 41 | 4119 | 79 | ||
XM_017015813.1 | XP_016871302.1 | 40 - 41 | 4141 | 79 | ||
XM_017015816.1 | XP_016871305.1 | 23 - 24 | 2389 | 79 | ||
GenBank mRNA | BC146857.1 | - | 40 - 41 | 4121 | 9056785 | 79 |
BX648342.1 | - | 12 - 13 | 2256 | 79 | ||
D50857.1 | - | 40 - 41 | 4082 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |