Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
TC01001738.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
ENST00000367049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
ENST00000367051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
ENST00000367052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
ENST00000367053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
ENST00000400960.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
NM_000573.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027229.hg.1 |
NM_000651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367052.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000400960.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000534202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
NM_000573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
NM_000651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367049.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367051.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367052.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000367053.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000400960.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
ENST00000534202.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
NM_000573.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
NM_000651.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184060.hg.1 |
TC0100011414.hg.1 |
Clariom D TC |