Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016939.hg.1 |
TC09001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016939.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016939.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000415441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000415441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000415441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000415441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010656.hg.1 |
ENST00000424473.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173853.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173853.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173853.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173853.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173853.hg.1 |
TC0900010865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
ENST00000375337.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010865.hg.1 |
NM_003837.3 |
Refseq |