Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
TC02000567.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
ENST00000309988.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008658.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008661.hg.1 |
TC02000567.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008661.hg.1 |
ENST00000309988.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008661.hg.1 |
ENST00000353004.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008661.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008661.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
TC02000567.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
ENST00000309988.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
ENST00000353004.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
ENST00000354078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02008659.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158474.hg.1 |
TC0200008452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158475.hg.1 |
TC0200008452.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000349807.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000354078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
ENST00000489399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008452.hg.1 |
NM_022440.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
ENST00000309988.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
ENST00000353004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
NM_002371.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
NM_022438.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200158477.hg.1 |
TC0200008452.hg.1 |
Clariom D TC |