Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009209.hg.1 |
ENST00000580897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009209.hg.1 |
ENST00000580897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009209.hg.1 |
ENST00000580897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026787.hg.1 |
ENST00000357736.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026787.hg.1 |
ENST00000392366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026787.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026787.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026787.hg.1 |
TC17001965.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026792.hg.1 |
ENST00000357736.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026792.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026792.hg.1 |
TC17001965.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026788.hg.1 |
ENST00000357736.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026788.hg.1 |
ENST00000392366.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026788.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026788.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026788.hg.1 |
TC17001965.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208326.hg.1 |
TC1700012483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208324.hg.1 |
TC1700012483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208325.hg.1 |
TC1700012483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208323.hg.1 |
TC1700012483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208327.hg.1 |
TC1700012483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700208333.hg.1 |
TC1700012483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009209.hg.1 |
ENST00000580897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009209.hg.1 |
ENST00000580897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009209.hg.1 |
ENST00000580897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000357736.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000392366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
ENST00000574686.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_002359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012483.hg.1 |
NM_032711.3 |
Refseq |