Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003509.2 | NP_003500.1 | 1 | 416 | 64 | |
GenBank mRNA | BC112254.1 | - | 1 | 494 | 8327722 | 64 |
BC112256.1 | - | 1 | 494 | 8327724 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630013 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 32521 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 25331 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3360216 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3393135 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2314 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P200168 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P122573 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P122573 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28171 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9010 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3360216 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3360216 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820153 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P200168 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P122573 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P122573 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P200168 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P122573 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P200168 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 228 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 62328 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 13821 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P200168 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630060 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15796474 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630051 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630029 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630012 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630048 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630038 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630057 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630063 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630047 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630024 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630025 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630030 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630011 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630042 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630040 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P200168 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003509_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117581 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117584 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8117582 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8117583 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8117580 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 214542_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11715219_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11715219_s_at | - | - |
Affymetrix Human Genome U219 Array | 11759252_at | - | - |
Affymetrix Human Genome U219 Array | 11759253_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11759253_x_at | - | - |
Affymetrix Human Genome U219 Array | 11759252_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR06002218.hg.1 | TC06004063.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06002218.hg.1 | ENST00000358739.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06002218.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007374.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007374.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007374.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007374.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007374.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149982.hg.1 | TC0600007374.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600149984.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149984.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149984.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149984.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149984.hg.1 | TC0600007374.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600149985.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149985.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149985.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149985.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149985.hg.1 | TC0600007374.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600007374.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007374.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007374.hg.1 | ENST00000358739.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007374.hg.1 | NM_003509.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007374.hg.1 | NM_003509.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 208583_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 214554_at | - | - |
Human Genome U133 Plus 2.0 Array | 208583_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 214554_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 207156_at | - | - |
Human Genome U133 Plus 2.0 Array | 214542_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 207156_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 214542_x_at | - | - |
Human Genome U133A 2.0 Array | 214554_PM_at | - | - |
Human Genome U133A 2.0 Array | 214554_at | - | - |
Human Genome U133A 2.0 Array | 214542_x_at | - | - |
Human Genome U133A 2.0 Array | 214542_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 207156_at | - | - |
Human Genome U133A 2.0 Array | 208583_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 207156_PM_at | - | - |
Human Genome U133A 2.0 Array | 208583_x_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003509_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw574958 | - | - |
Human Genome U95 Set | 34157_PM_f_at | - | - |
Human Genome U95 Set | 34156_i_at | - | - |
Human Genome U95 Set | 285_g_at | - | - |
Human Genome U95 Set | 285_PM_g_at | - | - |
Human Genome U95 Set | 31768_at | - | - |
Human Genome U95 Set | 31768_PM_at | - | - |
Human Genome U95 Set | 284_PM_at | - | - |
Human Genome U95 Set | 34156_PM_i_at | - | - |
Human Genome U95 Set | 284_at | - | - |
Human Genome U95 Set | 34157_f_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 214542_x_at10 | - | - |
Human Genome U133 Set | 214554_at | - | - |
Human Genome U133 Set | 214554_PM_at | - | - |
Human Genome U133 Set | 208583_PM_x_at | - | - |
Human Genome U133 Set | 214542_PM_x_at | - | - |
Human Genome U133 Set | 207156_PM_at | - | - |
Human Genome U133 Set | 208583_x_at | - | - |
Human Genome U133 Set | 214542_x_at | - | - |
Human Genome U133 Set | 207156_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031152 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031152 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1663453 | BC062305.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1663453 | BC112254.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15718713-S | BC062305.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15718713-S | NM_003509.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15718713-S | BC112254.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_15718713-S | BC112256.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_26131-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001240113 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001240113 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7040750 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_26131 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1663453 | BC112256.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1663453 | BC062305.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1663453 | NM_003509.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1663453 | BC112254.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1663453 | BC112256.1 | human_transcriptome |