Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001364.hg.1 |
TC12003223.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001364.hg.1 |
ENST00000539196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001364.hg.1 |
ENST00000539535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001364.hg.1 |
ENST00000541016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001364.hg.1 |
ENST00000546220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001364.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001365.hg.1 |
TC12003223.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001365.hg.1 |
ENST00000539196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001365.hg.1 |
ENST00000539535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001365.hg.1 |
ENST00000541016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001365.hg.1 |
ENST00000546220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001365.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000606112.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000606112.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
ENST00000620255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012591.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000261406.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000539196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000599672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000607161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
ENST00000611981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
NM_006331.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199060.hg.1 |
TC1200012591.hg.1 |
Clariom D TC |