Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026691.hg.1 |
TC03001465.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026691.hg.1 |
ENST00000296292.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026691.hg.1 |
ENST00000394738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026691.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000467048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
ENST00000471158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014001.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
ENST00000296292.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
ENST00000394738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
NM_052859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300208136.hg.1 |
TC0300014001.hg.1 |
Clariom D TC |