Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025701.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025701.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025701.hg.1 |
TC19001893.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025705.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025705.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025705.hg.1 |
TC19001893.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025704.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025704.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025704.hg.1 |
TC19001893.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025703.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025703.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025703.hg.1 |
TC19001893.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900197792.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197792.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197792.hg.1 |
TC1900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197792.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197792.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197796.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197796.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197796.hg.1 |
TC1900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197796.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197796.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197794.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197794.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197794.hg.1 |
TC1900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197794.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197794.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197795.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197795.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197795.hg.1 |
TC1900011548.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197795.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197795.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
ENST00000269834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011548.hg.1 |
NM_052882.1 |
Refseq |