Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021067.hg.1 |
TC05001544.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021067.hg.1 |
ENST00000307624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021067.hg.1 |
ENST00000508234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05021067.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500178312.hg.1 |
TC0500011296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000307624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000506440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000508234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
ENST00000513751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011296.hg.1 |
NM_130767.2 |
Refseq |