Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017831.hg.1 |
ENST00000247271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017831.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017831.hg.1 |
TC17001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017832.hg.1 |
ENST00000247271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017832.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017832.hg.1 |
TC17001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017833.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17017833.hg.1 |
TC17001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012383.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205933.hg.1 |
TC1700012383.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
ENST00000247271.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
ENST00000582029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
NM_002544.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205934.hg.1 |
TC1700012383.hg.1 |
Clariom D TC |