Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256327.1 | NP_001243256.1 | 26 - 27 | 5150 | 93 | |
NM_001256329.1 | NP_001243258.1 | 26 - 27 | 5048 | 93 | ||
NM_001256332.1 | NP_001243261.1 | 25 - 26 | 4979 | 93 | ||
NM_001256333.1 | NP_001243262.1 | 26 - 27 | 5150 | 93 | ||
NM_001256359.1 | NP_001243288.1 | 26 - 27 | 5069 | 93 | ||
NM_018896.4 | NP_061496.2 | 26 - 27 | 5150 | 93 | ||
NM_198382.2 | NP_938196.1 | 25 - 26 | 5081 | 93 | ||
NM_198383.2 | NP_938197.1 | 25 - 26 | 5081 | 93 | ||
NM_198384.2 | NP_938198.1 | 26 - 27 | 5150 | 93 | ||
NM_198385.2 | NP_938199.1 | 26 - 27 | 5150 | 93 | ||
NR_046054.1 | 26 - 27 | 5150 | 93 | |||
NR_046057.1 | 25 - 26 | 5081 | 93 | |||
XM_006722161.3 | XP_006722224.1 | 25 - 26 | 5169 | 93 | ||
GenBank mRNA | AF134986.1 | - | 26 - 27 | 4758 | 93 | |
AF227747.1 | - | 25 - 26 | 4689 | 93 | ||
AF227748.1 | - | 25 - 26 | 4689 | 93 | ||
AF227749.1 | - | 26 - 27 | 4758 | 93 | ||
AF227750.1 | - | 26 - 27 | 4758 | 93 | ||
BC110995.1 | - | 25 - 26 | 5081 | 5502027 | 93 | |
DQ494451.1 | - | 25 - 26 | 4689 | 93 | ||
DQ494452.1 | - | 25 - 26 | 4689 | 93 | ||
DQ494453.1 | - | 25 - 26 | 4689 | 93 | ||
DQ494461.1 | - | 26 - 27 | 4758 | 93 | ||
DQ494462.1 | - | 26 - 27 | 4758 | 93 | ||
DQ494463.1 | - | 26 - 27 | 4758 | 93 | ||
DQ494464.1 | - | 26 - 27 | 4758 | 93 | ||
DQ494465.1 | - | 26 - 27 | 4758 | 93 | ||
DQ494472.1 | - | 25 - 26 | 4587 | 93 | ||
DQ494474.1 | - | 26 - 27 | 4656 | 93 | ||
DQ494475.1 | - | 26 - 27 | 4677 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |