Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
TC22001497.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
ENST00000216271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
ENST00000349505.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
ENST00000448072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
ENST00000498366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
NM_001159286.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015259.hg.1 |
NM_032019.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
TC2200009360.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000216271.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000349505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000415993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000429374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000448072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000454936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000471375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000477814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000483222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000496235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000497952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000498366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000626012.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
NM_001159286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
NM_032019.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000216271.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000349505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000415993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000429374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000448072.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000454936.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000471375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000477814.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000483222.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000496235.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000497952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000498366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
ENST00000626012.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
NM_001159286.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170661.hg.1 |
NM_032019.5 |
Refseq |