Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009115.hg.1 |
ENST00000298440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009115.hg.1 |
ENST00000543457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009115.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009115.hg.1 |
TC13000813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009117.hg.1 |
ENST00000298440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009117.hg.1 |
ENST00000543457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009117.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009117.hg.1 |
TC13000813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009118.hg.1 |
ENST00000298440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009118.hg.1 |
ENST00000543457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009118.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009118.hg.1 |
TC13000813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009114.hg.1 |
ENST00000298440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009114.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009114.hg.1 |
TC13000813.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009111.hg.1 |
ENST00000298440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009111.hg.1 |
ENST00000543457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009111.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009111.hg.1 |
TC13000813.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163904.hg.1 |
TC1300009538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163898.hg.1 |
TC1300009538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163902.hg.1 |
TC1300009538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000541518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009538.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163896.hg.1 |
TC1300009538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163901.hg.1 |
TC1300009538.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
ENST00000298440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
ENST00000541038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
ENST00000543457.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
NM_080818.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300163905.hg.1 |
TC1300009538.hg.1 |
Clariom D TC |