Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014685.hg.1 |
ENST00000305738.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014685.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014685.hg.1 |
TC11001161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170844.hg.1 |
TC1100009426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000305738.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000437148.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
ENST00000525969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009426.hg.1 |
NM_138294.2 |
Refseq |