Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024546.hg.1 |
TC05003428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024546.hg.1 |
ENST00000503143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024546.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024544.hg.1 |
TC05003428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024544.hg.1 |
ENST00000503143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024544.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024545.hg.1 |
TC05003428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024545.hg.1 |
ENST00000503143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024545.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024548.hg.1 |
TC05003428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024548.hg.1 |
ENST00000503143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024548.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024547.hg.1 |
TC05003428.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024547.hg.1 |
ENST00000503143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024547.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196739.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196739.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196739.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196739.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196739.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500196735.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196735.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196735.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196735.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196735.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500196733.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196733.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196733.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196733.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196733.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500196734.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196734.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196734.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196734.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196734.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500196737.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196737.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196737.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196737.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196737.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500196736.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196736.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196736.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196736.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196736.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500196738.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196738.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196738.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196738.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196738.hg.1 |
TC0500013363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
ENST00000503143.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013363.hg.1 |
NM_130848.2 |
Refseq |