Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000466402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000466402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
ENST00000466402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013491.hg.1 |
TC09001028.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013491.hg.1 |
ENST00000361264.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013491.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013491.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167740.hg.1 |
TC0900009882.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000361264.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000396990.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000450964.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000463286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000466402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000466402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
ENST00000466402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009882.hg.1 |
NM_015397.3 |
Refseq |