Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000490148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000490148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
ENST00000490148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001417.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001417.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001417.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001417.hg.1 |
TC20000105.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
TC2000006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146459.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000246081.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000486129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000490148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000490148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
ENST00000490148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006782.hg.1 |
NM_020157.3 |
Refseq |