Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006637.hg.1 |
TC01006316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006637.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006637.hg.1 |
ENST00000373510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006637.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006633.hg.1 |
TC01006316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006633.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006635.hg.1 |
TC01006316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006635.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006635.hg.1 |
ENST00000373510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006635.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006638.hg.1 |
TC01006316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006638.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006638.hg.1 |
ENST00000373510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006638.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006636.hg.1 |
TC01006316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006636.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006636.hg.1 |
ENST00000373510.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01006636.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100237645.hg.1 |
TC0100018209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000316459.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
ENST00000373510.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001040441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NM_001291496.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018209.hg.1 |
NR_111980.1 |
Refseq |