Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000278499.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000416495.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000536441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000537480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000537480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000537480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000537480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
ENST00000542176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_001271594.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012019.hg.1 |
NM_144665.3 |
Refseq |