Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009456.hg.1 |
TC02004990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009456.hg.1 |
ENST00000302513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009456.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009456.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009457.hg.1 |
TC02004990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009457.hg.1 |
ENST00000302513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009457.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009457.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009452.hg.1 |
TC02004990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009452.hg.1 |
ENST00000302513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009452.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009452.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009454.hg.1 |
TC02004990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009454.hg.1 |
ENST00000302513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009454.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009454.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009449.hg.1 |
TC02004990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009449.hg.1 |
ENST00000302513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009449.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009450.hg.1 |
TC02004990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009450.hg.1 |
ENST00000302513.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009450.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02009450.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222732.hg.1 |
TC0200016497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222737.hg.1 |
TC0200016497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222730.hg.1 |
TC0200016497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222734.hg.1 |
TC0200016497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222729.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222729.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222729.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222729.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222729.hg.1 |
TC0200016497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222738.hg.1 |
TC0200016497.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000302513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000409684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
ENST00000496794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016497.hg.1 |
NM_144706.3 |
Refseq |