Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006689.hg.1 |
ENST00000379631.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006689.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006689.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006689.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006689.hg.1 |
TC13000583.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158496.hg.1 |
TC1300008659.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000379631.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
ENST00000423210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_001144033.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008659.hg.1 |
NM_145286.2 |
Refseq |