Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000406696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000406696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000406696.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000370836.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000406696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000406696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000406696.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
ENST00000521489.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_001077188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010840.hg.1 |
NM_147175.3 |
Refseq |