Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002851.hg.1 |
ENST00000397942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002851.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002851.hg.1 |
TC18000242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011815.hg.1 |
ENST00000528055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002852.hg.1 |
ENST00000397942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002852.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002852.hg.1 |
TC18000242.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
TC1800007620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152204.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007620.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
TC1800007620.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
ENST00000397942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
ENST00000578377.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
ENST00000582322.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152203.hg.1 |
NM_004232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011815.hg.1 |
ENST00000528055.1 |
Ensembl |