Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022572.hg.1 |
TC01001452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022572.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022572.hg.1 |
ENST00000367876.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022572.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022571.hg.1 |
TC01001452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022571.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022571.hg.1 |
ENST00000367876.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022571.hg.1 |
ENST00000537173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022571.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022570.hg.1 |
TC01001452.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022570.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022570.hg.1 |
ENST00000367876.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022570.hg.1 |
ENST00000537173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022570.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177132.hg.1 |
TC0100010486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177131.hg.1 |
TC0100010486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |