Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7960945 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7960946 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7960944 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7960943 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7960942 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1555040_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015712.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015712.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015712.hg.1 |
TC12001186.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015713.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015713.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015713.hg.1 |
TC12001186.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200174046.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174046.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174046.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174046.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174046.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174043.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174043.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174043.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174043.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174043.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174045.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174045.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174045.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174045.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174045.hg.1 |
TC1200009846.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
ENST00000538094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009846.hg.1 |
NR_034140.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1555040_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1555040_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555040_at10 |
- |
- |