Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004656.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004656.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004656.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004656.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004656.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004656.hg.1 |
TC21000411.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004662.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004662.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004662.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004662.hg.1 |
TC21000411.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004657.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004657.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004657.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004657.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004657.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004657.hg.1 |
TC21000411.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000314399.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
ENST00000617313.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007999.hg.1 |
NM_017833.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153916.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153916.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153916.hg.1 |
TC2100007999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153916.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153916.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
NM_001040192.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
TC2100007999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
ENST00000381947.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
ENST00000402202.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153915.hg.1 |
NM_001040192.2 |
Refseq |