Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008512.hg.1 |
ENST00000320460.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008512.hg.1 |
ENST00000378692.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008512.hg.1 |
ENST00000441218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008512.hg.1 |
ENST00000550916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008512.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12008512.hg.1 |
TC12000641.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200160455.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160455.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160455.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160455.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160455.hg.1 |
TC1200008223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200160457.hg.1 |
TC1200008223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000320460.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000378692.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000547164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
ENST00000550916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_001270396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NM_152436.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008223.hg.1 |
NR_072995.1 |
Refseq |