Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170733.3 | NP_733929.1 | 1 - 2 | 333 | 116 | |
GenBank mRNA | AB038672.1 | - | 1 - 2 | 215 | 116 | |
AY054397.2 | - | 1 - 2 | 215 | 116 | ||
BC029795.1 | - | 1 - 2 | 306 | 5193877 | 116 | |
EF674521.1 | - | 1 - 2 | 333 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Homo sapiens Human Growth Factors Inventoried Human_Growth_Factors Probe spans exons |