Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009814.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009814.hg.1 |
ENST00000447138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009814.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009814.hg.1 |
TC20000726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009815.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009815.hg.1 |
ENST00000447138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009815.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009815.hg.1 |
TC20000726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009816.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009816.hg.1 |
ENST00000447138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009816.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009816.hg.1 |
TC20000726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009818.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009818.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009818.hg.1 |
TC20000726.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160958.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160960.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160960.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160960.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160960.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160960.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160961.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160961.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160961.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160961.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160961.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160957.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160959.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160959.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160959.hg.1 |
TC2000008721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160959.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160959.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000217456.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
ENST00000451442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008721.hg.1 |
NM_020531.2 |
Refseq |