Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002950.hg.1 |
TC01000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002950.hg.1 |
ENST00000400796.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002950.hg.1 |
ENST00000434578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002950.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002950.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
TC01000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
ENST00000303840.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
ENST00000376008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
ENST00000400796.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
ENST00000434578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002952.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148513.hg.1 |
TC0100006977.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148511.hg.1 |
TC0100006977.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376008.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000376014.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000400796.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000428417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000434578.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
ENST00000451326.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136217.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_001136218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006977.hg.1 |
NM_018022.2 |
Refseq |