Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224499.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224499.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224499.hg.1 |
TC0100016363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016363.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
ENST00000303469.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
ENST00000310392.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
ENST00000454472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
NM_033418.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224498.hg.1 |
TC0100016363.hg.1 |
Clariom D TC |