Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000508882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000508882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000508882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002099.hg.1 |
TC05000179.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002099.hg.1 |
ENST00000274276.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002099.hg.1 |
ENST00000502536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002099.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002099.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148726.hg.1 |
TC0500007201.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000274276.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000502536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000508882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000508882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000508882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000509237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
ENST00000513831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_001168355.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007201.hg.1 |
NM_003999.2 |
Refseq |