Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023090.hg.1 |
TC09001775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023090.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023090.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023090.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023092.hg.1 |
TC09001775.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023092.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023092.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09023092.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012076.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183254.hg.1 |
TC0900012076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
ENST00000298585.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
ENST00000471957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
NM_138462.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183256.hg.1 |
TC0900012076.hg.1 |
Clariom D TC |