Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011330.hg.1 |
ENST00000220514.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011330.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15011330.hg.1 |
TC15001248.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500166272.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500166272.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500166272.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500166272.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500166272.hg.1 |
TC1500009148.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000220514.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
ENST00000560640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009148.hg.1 |
NM_007280.1 |
Refseq |