Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011437.hg.1 |
TC01000720.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011437.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011437.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011437.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008588.hg.1 |
NR_002771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160923.hg.1 |
TC0100008588.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371086.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000371088.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000460678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000461039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000480886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000493605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
ENST00000496956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NM_032437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008588.hg.1 |
NR_002771.1 |
Refseq |