Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026831.hg.1 |
ENST00000308366.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026831.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026831.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026831.hg.1 |
TC12002042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026829.hg.1 |
ENST00000308366.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026829.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026829.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026829.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026829.hg.1 |
TC12002042.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194810.hg.1 |
TC1200012158.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012158.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194811.hg.1 |
TC1200012158.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
ENST00000308366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
ENST00000423423.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
ENST00000549767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
NM_000928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194808.hg.1 |
TC1200012158.hg.1 |
Clariom D TC |