Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002681.hg.1 |
TC01000156.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002681.hg.1 |
ENST00000356315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002681.hg.1 |
ENST00000358136.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002681.hg.1 |
ENST00000496628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002681.hg.1 |
NM_015378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002681.hg.1 |
NM_018156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000011700.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000543710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000613099.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000620676.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
NM_015378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
NM_018156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000011700.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000543710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000613099.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
ENST00000620676.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
NM_015378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
NM_018156.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148039.hg.1 |
TC0100006884.hg.1 |
Clariom D TC |