Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000492603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
ENST00000492603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004372.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004372.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004372.hg.1 |
TC21000397.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000480196.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000492603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
ENST00000492603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007952.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153498.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153498.hg.1 |
NM_014825.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153498.hg.1 |
TC2100007952.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153498.hg.1 |
ENST00000382751.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153498.hg.1 |
NM_014825.2 |
Refseq |