Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006915.hg.1 |
ENST00000222345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006915.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006915.hg.1 |
TC19000521.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000476317.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000476317.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000476317.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000595982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000599644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000599644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000600919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000600919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000600919.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
TC1900007988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900159810.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000222345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000476317.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000476317.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000476317.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000594553.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595495.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000595982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000596403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000599644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000599644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000600919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000600919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000600919.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
ENST00000601881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007988.hg.1 |
NM_015073.2 |
Refseq |