Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000262320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000354866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000457798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000461023.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
ENST00000481769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_003502.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008938.hg.1 |
NM_181050.2 |
Refseq |