Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018572.hg.1 |
TC09001492.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018572.hg.1 |
ENST00000374238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018572.hg.1 |
ENST00000374242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018572.hg.1 |
ENST00000497712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018572.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
TC09001492.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
ENST00000374236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
ENST00000374238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
ENST00000374242.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
ENST00000497712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018573.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176529.hg.1 |
TC0900011236.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011236.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000374234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000374236.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000374242.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000476599.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000481146.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
ENST00000497712.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
NM_021218.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176530.hg.1 |
TC0900011236.hg.1 |
Clariom D TC |