Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012718.hg.1 |
TC06001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012718.hg.1 |
ENST00000367467.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012718.hg.1 |
ENST00000535767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012718.hg.1 |
ENST00000537769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012718.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012717.hg.1 |
TC06001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012717.hg.1 |
ENST00000367467.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012717.hg.1 |
ENST00000535767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012717.hg.1 |
ENST00000537769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012717.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166979.hg.1 |
TC0600009772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166980.hg.1 |
TC0600009772.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |