Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022373.hg.1 |
ENST00000311915.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022373.hg.1 |
ENST00000398055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022373.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022373.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022373.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022373.hg.1 |
TC12001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022370.hg.1 |
ENST00000311915.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022370.hg.1 |
ENST00000398055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022370.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022370.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022370.hg.1 |
TC12001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022372.hg.1 |
ENST00000311915.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022372.hg.1 |
ENST00000398055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022372.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022372.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022372.hg.1 |
TC12001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022376.hg.1 |
ENST00000398055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022376.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022376.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022376.hg.1 |
TC12001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186141.hg.1 |
TC1200011081.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |